3-(Trifluoromethylthio)phenyl isocyanate 95%

Supplier: Apollo Scientific

PC0086-1G PC0086-250MG
APOSPC0086-1GEA 870 EUR
APOSPC0086-1G APOSPC0086-250MG
3-(Trifluoromethylthio)phenyl isocyanate 95%
3-(Trifluoromethylthio)phenyl isocyanate

May form small amount of sediment

Formula: C₈H₄F₃NOS
Boiling Pt: 45 °C (0,01 mm Hg)
Density: 1,365 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00673057
CAS Number: 55225-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR