4-Bromo-5-fluoro-2-(2,2,2-trifluoroethoxy)aniline

Supplier: Apollo Scientific
Warning

PC01490-1G PC01490-5G PC01490-250MG
APOSPC01490-1GEA 592 EUR
APOSPC01490-1G APOSPC01490-5G APOSPC01490-250MG
4-Bromo-5-fluoro-2-(2,2,2-trifluoroethoxy)aniline
4-Bromo-5-fluoro-2-(2,2,2-trifluoroethoxy)aniline
Formula: C₈H₆BrF₄NO
MW: 288,037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28155018
CAS Number: 1602345-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR