Phenyl 2-fluoro-5-nitrobenzoate

Supplier: Apollo Scientific

Synonyms: 4-Fluoro-3-(phenoxycarbonyl)nitrobenzene

PC01653-25G PC01653-5G
APOSPC01653-25GEA 480 EUR
APOSPC01653-25G APOSPC01653-5G
Phenyl 2-fluoro-5-nitrobenzoate
Phenyl 2-fluoro-5-nitrobenzoate
Formula: C₁₃H₈FNO₄
MW: 261,205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16170460
CAS Number: 1257535-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR