2-Amino-5-bromo-4-fluorophenol

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromo-5-fluoro-2-hydroxyaniline

PC01680-1G PC01680-250MG PC01680-5G
APOSPC01680-1GEA 161 EUR
APOSPC01680-1G APOSPC01680-250MG APOSPC01680-5G
2-Amino-5-bromo-4-fluorophenol
2-Amino-5-bromo-4-fluorophenol
Formula: C₆H₅BrFNO
MW: 206,012 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24640741
CAS Number: 1037364-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR