Hexakis(1H,1H,5H-perfluoropentoxy)phosphazene 95%

Supplier: Apollo Scientific
Warning

PC0233-1G PC0233-250MG
APOSPC0233-1GEA 596 EUR
APOSPC0233-1G APOSPC0233-250MG
Hexakis(1H,1H,5H-perfluoropentoxy)phosphazene 95%
Hexakis(1H,1H,5H-perfluoropentoxy)phosphazene
Formula: C₃₀H₁₈F₄₈N₃O₆P₃
MW: 1521,325 g/mol
Boiling Pt: 203…207 °C (0,25 mm Hg)
Melting Pt: 14…16 °C
Storage Temperature: Ambient
MDL Number: MFCD00798142
CAS Number: 16059-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR