4-Methyl-3-(trifluoromethyl)benzyl alcohol

Supplier: Apollo Scientific
Warning

PC0478-5G PC0478-250MG
APOSPC0478-5GEA 458 EUR
APOSPC0478-5G APOSPC0478-250MG
4-Methyl-3-(trifluoromethyl)benzyl alcohol
4-Methyl-3-(trifluoromethyl)benzyl alcohol
Formula: C₉H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD01631607
CAS Number: 261952-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR