2-Phenyl-1,1,1-trifluoropropan-2-ol 97%

Supplier: Apollo Scientific
Warning

PC0487-1G PC0487-25G PC0487-5G
APOSPC0487-1GEA 115 EUR
APOSPC0487-1G APOSPC0487-25G APOSPC0487-5G
2-Phenyl-1,1,1-trifluoropropan-2-ol 97%
2-Phenyl-1,1,1-trifluoropropan-2-ol
Formula: C₉H₉F₃O
MW: 190,162 g/mol
Boiling Pt: 81…83 °C (3 mm Hg)
Density: 1,251 g/cm³ (25 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD07366525
CAS Number: 426-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR