4-(Trifluoromethyl)cinnamamide 98%

Supplier: Apollo Scientific
Warning

PC0552-1G PC0552-5G
APOSPC0552-1GEA 23.2 EUR
APOSPC0552-1G APOSPC0552-5G
4-(Trifluoromethyl)cinnamamide 98%
4-(Trifluoromethyl)cinnamamide
Formula: C₁₀H₈F₃NO
MW: 215.172 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03092988
CAS Number: 115093-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR