(1S)-1-[2-(Trifluoromethyl)phenyl]ethylamine 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(S)-α-Methyl-2-(trifluoromethyl)benzylamine, 2-[(1S)-1-Aminoethyl]benzotrifluoride
PC0598-100MG
PC0598-250MG
APOSPC0598-100MGEA
263
EUR
APOSPC0598-100MG
APOSPC0598-250MG
(1S)-1-[2-(Trifluoromethyl)phenyl]ethylamine 98%
(1S)-1-[2-(Trifluoromethyl)phenyl]ethylamine
Formula:
C₉H₁₀F₃N MW: 189,178 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03093003 CAS Number: 127733-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...