4-Fluoro-4'-nitrochalcone

Supplier: Apollo Scientific
Warning

Synonyms: 3-(4-Fluorophenyl)-1-(4-nitrophenyl)prop-2-en-1-one, 4-[3-(4-Fluorophenyl)acryloyl]nitrobenzene

PC0639-1G PC0639-5G
APOSPC0639-1GEA 140 EUR
APOSPC0639-1G APOSPC0639-5G
4-Fluoro-4'-nitrochalcone
4-Fluoro-4'-nitrochalcone
Formula: C₁₅H₁₀FNO₃
MW: 271,243 g/mol
Melting Pt: 203…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00024742
CAS Number: 2805-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR