3-Fluoro-2-iodoanisole
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Fluoro-6-methoxyiodobenzene, 3-Fluoro-2-iodophenyl methyl ether, 1-Fluoro-2-iodo-3-methoxybenzene
PC0873-1G
PC0873-25G
PC0873-5G
APOSPC0873-1GEA
40.6
EUR
APOSPC0873-1G
APOSPC0873-25G
APOSPC0873-5G
3-Fluoro-2-iodoanisole
3-Fluoro-2-iodoanisole
Formula:
C₇H₆FIO MW: 252,025 g/mol Boiling Pt: 240 °C (746 mm Hg) Density: 1,859 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD03788539 CAS Number: 7079-54-1 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...