1,1,1-Trifluorobutan-2-ol 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Hydroxy-1,1,1-trifluorobutane, 1-(Trifluoromethyl)propan-1-ol
PC0984-1G
PC0984-5G
APOSPC0984-1GEA
198
EUR
APOSPC0984-1G
APOSPC0984-5G
1,1,1-Trifluorobutan-2-ol 97%
1,1,1-Trifluorobutan-2-ol
Formula:
C₄H₇F₃O Boiling Pt: 90,8…91.8 °C (752 mm Hg) Density: 1,166 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00462509 CAS Number: 431-36-7 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...