2-Sulphanyl-4-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Mercapto-4-(trifluoromethyl)pyridine, 4-(Trifluoromethyl)pyridine-2-thiol, 2-Sulphanyl-α,α,α-trifluoro-4-picoline
PC103684-1G
PC103684-250MG
APOSPC103684-1GEA
299
EUR
APOSPC103684-1G
APOSPC103684-250MG
2-Sulphanyl-4-(trifluoromethyl)pyridine
2-Sulphanyl-4-(trifluoromethyl)pyridine
Formula:
C₆H₄F₃NS MW: 179,163 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09881227 CAS Number: 121307-79-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...