2-Fluoro-6-iodoaniline 99%

Supplier: Apollo Scientific
Warning

PC10400-1G PC10400-5G PC10400-25G
APOSPC10400-1GEA 98.9 EUR
APOSPC10400-1G APOSPC10400-5G APOSPC10400-25G
2-Fluoro-6-iodoaniline 99%
2-Fluoro-6-iodoaniline
Formula: C₆H₅FIN
MW: 237,013 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368681
CAS Number: 886762-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR