Dibromomethylpentafluorobenzene

Supplier: Apollo Scientific
Warning

PC10599-1G PC10599-5G
APOSPC10599-1GEA 139 EUR
APOSPC10599-1G APOSPC10599-5G
Dibromomethylpentafluorobenzene
Dibromomethylpentafluorobenzene
Formula: C₇HBr₂F₅
Storage Temperature: Ambient
MDL Number: MFCD07368710
CAS Number: 887266-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR