4-(1,1,1,3,3,3-Hexafluoro-2-hydroxyprop-2-yl)phenol 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
α,α-Bis(trifluoromethyl)-4-hydroxybenzyl alcohol, 4-(1,1,1,3,3,3-Hexafluoro-2-hydroxypropan-2-yl)phenol, 1,1,1,3,3,3-Hexafluoro-2-(4-hydroxyphenyl)propan-2-ol
PC10607-1G
PC10607-5G
APOSPC10607-1GEA
401
EUR
APOSPC10607-1G
APOSPC10607-5G
4-(1,1,1,3,3,3-Hexafluoro-2-hydroxyprop-2-yl)phenol 95%
4-(1,1,1,3,3,3-Hexafluoro-2-hydroxyprop-2-yl)phenol
Formula:
C₉H₆F₆O₂ Melting Pt: 122,5…123.5 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD01050148 CAS Number: 836-79-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...