4-Fluoro-2-iodophenol 95%

Supplier: Apollo Scientific
Warning

PC10631-1G PC10631-5G PC10631-25G
APOSPC10631-1GEA 14.7 EUR
APOSPC10631-1G APOSPC10631-5G APOSPC10631-25G
4-Fluoro-2-iodophenol 95%
4-Fluoro-2-iodophenol
Formula: C₆H₄FIO
MW: 237,998 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15527539
CAS Number: 2713-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR