3-Fluoro-6-methylpyridine-2-carboxylic acid

Supplier: Apollo Scientific
Warning

Synonyms: 2-Carboxy-3-fluoro-6-methylpyridine, 3-Fluoro-6-methylpicolinic acid

PC10658-1G PC10658-250MG
APOSPC10658-1GEA 560 EUR
APOSPC10658-1G APOSPC10658-250MG
3-Fluoro-6-methylpyridine-2-carboxylic acid
3-Fluoro-6-methylpyridine-2-carboxylic acid
Formula: C₇H₆FNO₂
MW: 155,126 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17214194
CAS Number: 1256806-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR