4-[(Trifluoromethyl)sulphonyl]benzene-1,2-diamine 95%
Supplier: Apollo Scientific
|
Warning
|
PC10662-1G
PC10662-250MG
PC10662-5G
APOSPC10662-1GEA
209
EUR
APOSPC10662-1G
APOSPC10662-250MG
APOSPC10662-5G
4-[(Trifluoromethyl)sulphonyl]benzene-1,2-diamine 95%
4-[(Trifluoromethyl)sulphonyl]benzene-1,2-diamine
Formula:
C₇H₇F₃N₂O₂S Melting Pt: 109…111 °C Storage Temperature: Ambient |
MDL Number:
MFCD00491700 CAS Number: 2355-16-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...