3-Amino-4-fluorobenzo[b]thiophene-2-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
PC110001-1G
PC110001-250MG
APOSPC110001-1GEA
499
EUR
APOSPC110001-1G
APOSPC110001-250MG
3-Amino-4-fluorobenzo[b]thiophene-2-carboxylic acid
3-Amino-4-fluorobenzo[b]thiophene-2-carboxylic acid
Formula:
C₉H₆FNO₂S MW: 211,213 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05664782 CAS Number: 874800-63-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...