3-Amino-4-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid

Supplier: Apollo Scientific
Warning

PC110014-1G PC110014-250MG
APOSPC110014-1GEA 662 EUR
APOSPC110014-1G APOSPC110014-250MG
3-Amino-4-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid
3-Amino-4-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid
Formula: C₁₀H₆F₃NO₂S
MW: 261,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29058792
CAS Number: 1648807-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR