5-Fluoro-6-hydroxypyridine-2-carbonitrile
Supplier: Apollo Scientific
|
Warning
|
PC110024-1G
PC110024-250MG
PC110024-5G
APOSPC110024-1GEA
173
EUR
APOSPC110024-1G
APOSPC110024-250MG
APOSPC110024-5G
5-Fluoro-6-hydroxypyridine-2-carbonitrile
5-Fluoro-6-hydroxypyridine-2-carbonitrile
Formula:
C₆H₃FN₂O MW: 138,099 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28768731 CAS Number: 1239510-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...