3-(4-Fluorophenyl)prop-2-yn-1-ol 97%

Supplier: Apollo Scientific
Warning

PC11156-1G PC11156-250MG PC11156-5G
APOSPC11156-1GEA 176 EUR
APOSPC11156-1G APOSPC11156-250MG APOSPC11156-5G
3-(4-Fluorophenyl)prop-2-yn-1-ol 97%
3-(4-Fluorophenyl)prop-2-yn-1-ol
Formula: C₉H₇FO
Melting Pt: 32…33 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039232
CAS Number: 80151-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR