1-[4-(Trifluoromethoxy)benzyl]piperazine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-[(Piperazin-1-yl)methyl]-4-(trifluoromethoxy)benzene
PC1414-1G
PC1414-25G
PC1414-5G
APOSPC1414-1GEA
46.6
EUR
APOSPC1414-1G
APOSPC1414-25G
APOSPC1414-5G
1-[4-(Trifluoromethoxy)benzyl]piperazine
1-[4-(Trifluoromethoxy)benzyl]piperazine
Formula:
C₁₂H₁₅F₃N₂O MW: 260,256 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07368745 CAS Number: 340759-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...