3-Fluoro-5-isopropoxybenzeneboronic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Fluoro-5-isopropoxyphenylboronic acid, (3-Fluoro-5-isopropoxyphenyl)boronic acid
PC1609-1G
PC1609-5G
PC1609-25G
APOSPC1609-1GEA
91.6
EUR
APOSPC1609-1G
APOSPC1609-5G
APOSPC1609-25G
3-Fluoro-5-isopropoxybenzeneboronic acid 98%
3-Fluoro-5-isopropoxybenzeneboronic acid
Formula:
C₉H₁₂BFO₃ MW: 197,999 g/mol Melting Pt: 136…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD07363781 CAS Number: 1195945-65-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...