Graphite, fluorinated, polymer
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Carbon monofluoride
PC1649L-1G
PC1649L-5G
APOSPC1649L-1GEA
62.5
EUR
APOSPC1649L-1G
APOSPC1649L-5G
Graphite, fluorinated, polymer
Graphite, fluorinated, polymer
MW:
31.009 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00135559 CAS Number: 51311-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...