3-(2,2,2-Trifluoroethoxy)propionitrile

Supplier: Apollo Scientific
Warning

PC1664-25G PC1664-5G
APOSPC1664-25GEA 329 EUR
APOSPC1664-25G APOSPC1664-5G
3-(2,2,2-Trifluoroethoxy)propionitrile
3-(2,2,2-Trifluoroethoxy)propionitrile
Formula: C₅H₆F₃NO
Boiling Pt: 186 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07784292
CAS Number: 272128-06-0
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR