Ethyl 6-fluoro-2-methylquinoline-3-carboxylate

Supplier: Apollo Scientific

PC1804-1G PC1804-5G
APOSPC1804-1GEA 224 EUR
APOSPC1804-1G APOSPC1804-5G
Ethyl 6-fluoro-2-methylquinoline-3-carboxylate
Ethyl 6-fluoro-2-methylquinoline-3-carboxylate
Formula: C₁₃H₁₂FNO₂
MW: 233,238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03541053
CAS Number: 282540-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR