2,3,5-Trichloro-4-trifluoromethyl pyridine

Supplier: Apollo Scientific

PC200033-1G PC200033-500MG
APOSPC200033-1GEA 423 EUR
APOSPC200033-1G APOSPC200033-500MG
2,3,5-Trichloro-4-trifluoromethyl pyridine
2,3,5-Trichloro-4-trifluoromethyl pyridine
Formula: C₆HCl₃F₃N
MW: 250,433 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22683240
CAS Number: 89719-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR