7-Chloro-6-fluoro-1-methoxyisoquinoline

Supplier: Apollo Scientific

PC200044-1G PC200044-500MG
APOSPC200044-1GEA 538 EUR
APOSPC200044-1G APOSPC200044-500MG
7-Chloro-6-fluoro-1-methoxyisoquinoline
7-Chloro-6-fluoro-1-methoxyisoquinoline
Formula: C₁₀H₇ClFNO
MDL Number: MFCD18374925
CAS Number: 1202006-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR