1-(2-Fluorophenyl)-4-oxocyclohexane-1-carbonitrile
Supplier: Apollo Scientific
PC200055-1G
PC200055-500MG
PC200055-5G
APOSPC200055-1GEA
538
EUR
APOSPC200055-1G
APOSPC200055-500MG
APOSPC200055-5G
1-(2-Fluorophenyl)-4-oxocyclohexane-1-carbonitrile
1-(2-Fluorophenyl)-4-oxocyclohexane-1-carbonitrile
Formula:
C₁₃H₁₂FNO MW: 217,239 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11849774 CAS Number: 179064-61-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...