2,5-Difluoro-3-methylbenzonitrile 98%
Supplier: Apollo Scientific
PC201002-1G
PC201002-25G
PC201002-5G
APOSPC201002-1GEA
43.9
EUR
APOSPC201002-1G
APOSPC201002-25G
APOSPC201002-5G
2,5-Difluoro-3-methylbenzonitrile 98%
2,5-Difluoro-3-methylbenzonitrile
Formula:
C₈H₅F₂N Storage Temperature: Ambient |
MDL Number:
MFCD09996954 CAS Number: 1003712-20-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...