3-Chloro-2-fluoroisonicotinaldehyde 95+%

Supplier: Apollo Scientific

PC201030-100MG PC201030-1G PC201030-250MG
APOSPC201030-100MGEA 49.7 EUR
APOSPC201030-100MG APOSPC201030-1G APOSPC201030-250MG
3-Chloro-2-fluoroisonicotinaldehyde 95+%
3-Chloro-2-fluoroisonicotinaldehyde
Formula: C₆H₃ClFNO
Storage Temperature: Ambient
MDL Number: MFCD12756634
CAS Number: 1149587-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR