5-Fluoro-1H-pyrrolo[2,3-c]pyridine 95+%
Supplier: Apollo Scientific
PC201044-100MG
PC201044-1G
PC201044-250MG
APOSPC201044-100MGEA
52.2
EUR
APOSPC201044-100MG
APOSPC201044-1G
APOSPC201044-250MG
5-Fluoro-1H-pyrrolo[2,3-c]pyridine 95+%
5-Fluoro-1H-pyrrolo[2,3-c]pyridine
Formula:
C₇H₅FN₂ MW: 136,126 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12962571 CAS Number: 1190319-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...