6-Chloro-4-(trifluoromethyl)nicotinonitrile 95%

Supplier: Apollo Scientific

PC201050-1G PC201050-250MG
APOSPC201050-1GEA 133 EUR
APOSPC201050-1G APOSPC201050-250MG
6-Chloro-4-(trifluoromethyl)nicotinonitrile 95%
6-Chloro-4-(trifluoromethyl)nicotinonitrile
Formula: C₇H₂ClF₃N₂
MW: 206,552 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15528775
CAS Number: 1201187-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR