4-(Trifluoromethyl)isoindoline hydrochloride 95+%
Supplier: Apollo Scientific
PC201053-100MG
PC201053-1G
PC201053-250MG
APOSPC201053-100MGEA
183
EUR
APOSPC201053-100MG
APOSPC201053-1G
APOSPC201053-250MG
4-(Trifluoromethyl)isoindoline hydrochloride 95+%
4-(Trifluoromethyl)isoindoline hydrochloride
Formula:
C₉H₈F₃N·ClH MW: 223,623 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10700141 CAS Number: 1203686-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...