3-Cyclopropyl-2-fluoropyridine 97%

Supplier: Apollo Scientific

PC201078-100MG PC201078-1G PC201078-250MG
APOSPC201078-100MGEA 177 EUR
APOSPC201078-100MG APOSPC201078-1G APOSPC201078-250MG
3-Cyclopropyl-2-fluoropyridine 97%
3-Cyclopropyl-2-fluoropyridine
Formula: C₈H₈FN
MW: 137,154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528324
CAS Number: 1227177-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR