3-Bromo-5-fluoroisonicotinonitrile 97%

Supplier: Apollo Scientific

PC201128-100MG PC201128-1G PC201128-250MG
APOSPC201128-100MGEA 82.9 EUR
APOSPC201128-100MG APOSPC201128-1G APOSPC201128-250MG
3-Bromo-5-fluoroisonicotinonitrile 97%
3-Bromo-5-fluoroisonicotinonitrile
Formula: C₆H₂BrFN₂
MW: 200,996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20524846
CAS Number: 1353636-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR