6-Fluoroquinolin-8-ol 95%
Supplier: Apollo Scientific
PC201129-100MG
PC201129-250MG
APOSPC201129-100MGEA
306
EUR
APOSPC201129-100MG
APOSPC201129-250MG
6-Fluoroquinolin-8-ol 95%
6-Fluoroquinolin-8-ol
Formula:
C₉H₆FNO MW: 163,148 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18414597 CAS Number: 135838-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...