2,6-Dichloro-5-fluoropyridin-3-amine 98%

Supplier: Apollo Scientific

PC201147-1G PC201147-5G PC201147-10G
APOSPC201147-1GEA 102 EUR
APOSPC201147-1G APOSPC201147-5G APOSPC201147-10G
2,6-Dichloro-5-fluoropyridin-3-amine 98%
2,6-Dichloro-5-fluoropyridin-3-amine
Formula: C₅H₃Cl₂FN₂
Storage Temperature: Ambient
MDL Number: MFCD11044255
CAS Number: 152840-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR