3-Bromo-4-chloro-2-fluoroaniline 97%

Supplier: Apollo Scientific

PC201149-100MG PC201149-1G PC201149-250MG
APOSPC201149-100MGEA 193 EUR
APOSPC201149-100MG APOSPC201149-1G APOSPC201149-250MG
3-Bromo-4-chloro-2-fluoroaniline 97%
3-Bromo-4-chloro-2-fluoroaniline
Formula: C₆H₄BrClFN
MW: 224,458 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29058770
CAS Number: 1539469-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR