Methyl 4-amino-3-(trifluoromethyl)benzoate 98%

Supplier: Apollo Scientific

PC201164-1G PC201164-5G
APOSPC201164-1GEA 95.7 EUR
APOSPC201164-1G APOSPC201164-5G
Methyl 4-amino-3-(trifluoromethyl)benzoate 98%
Methyl 4-amino-3-(trifluoromethyl)benzoate
Formula: C₉H₈F₃NO₂
MW: 219,161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226326
CAS Number: 167760-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR