(3-((Dimethylamino)methyl)-4-fluorophenyl)boronic acid 95+%
Supplier: Apollo Scientific
PC201169-100MG
PC201169-1G
PC201169-250MG
APOSPC201169-100MGEA
187
EUR
APOSPC201169-100MG
APOSPC201169-1G
APOSPC201169-250MG
(3-((Dimethylamino)methyl)-4-fluorophenyl)boronic acid 95+%
(3-((Dimethylamino)methyl)-4-fluorophenyl)boronic acid
Formula:
C₉H₁₃BFNO₂ MW: 197,014 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27935575 CAS Number: 1704063-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...