2-Fluoro-1,3-dimethyl-5-nitrobenzene 97%
Supplier: Apollo Scientific
|
Warning
|
PC201172-25G
PC201172-5G
PC201172-1G
APOSPC201172-25GEA
415
EUR
APOSPC201172-25G
APOSPC201172-5G
APOSPC201172-1G
2-Fluoro-1,3-dimethyl-5-nitrobenzene 97%
2-Fluoro-1,3-dimethyl-5-nitrobenzene
Formula:
C₈H₈FNO₂ MW: 169,153 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD22055006 CAS Number: 1736-85-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...