Methyl 7-fluoro-1H-indole-5-carboxylate 97%

Supplier: Apollo Scientific

PC201215-100MG PC201215-1G PC201215-250MG
APOSPC201215-100MGEA 235 EUR
APOSPC201215-100MG APOSPC201215-1G APOSPC201215-250MG
Methyl 7-fluoro-1H-indole-5-carboxylate 97%
Methyl 7-fluoro-1H-indole-5-carboxylate
Formula: C₁₀H₈FNO₂
MW: 193,174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06656840
CAS Number: 256935-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR