6-(Difluoromethoxy)pyridin-3-amine 95%

Supplier: Apollo Scientific
Warning

PC201228-1G PC201228-250MG PC201228-5G
APOSPC201228-1GEA 174 EUR
APOSPC201228-1G APOSPC201228-250MG APOSPC201228-5G
6-(Difluoromethoxy)pyridin-3-amine 95%
6-(Difluoromethoxy)pyridin-3-amine
Formula: C₆H₆F₂N₂O
MW: 160,121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847318
CAS Number: 317810-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR