2-Bromo-4-(trifluoromethyl)thiazole 97%

Supplier: Apollo Scientific

PC201251-1G PC201251-5G
APOSPC201251-1GEA 72.5 EUR
APOSPC201251-1G APOSPC201251-5G
2-Bromo-4-(trifluoromethyl)thiazole 97%
2-Bromo-4-(trifluoromethyl)thiazole
Formula: C₄HBrF₃NS
MW: 232,022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14702712
CAS Number: 41731-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR