5,7-Difluorochroman-4-one 95%
Supplier: Apollo Scientific
PC201320-250MG
PC201320-1G
PC201320-5G
APOSPC201320-250MGEA
132
EUR
APOSPC201320-250MG
APOSPC201320-1G
APOSPC201320-5G
5,7-Difluorochroman-4-one 95%
5,7-Difluorochroman-4-one
Formula:
C₉H₆F₂O₂ MW: 184,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11518474 CAS Number: 844648-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...