2-Fluoro-N,N-dimethylpyridin-4-amine 97%

Supplier: Apollo Scientific

PC201323-100MG PC201323-250MG PC201323-1G
APOSPC201323-100MGEA 91.6 EUR
APOSPC201323-100MG APOSPC201323-250MG APOSPC201323-1G
2-Fluoro-N,N-dimethylpyridin-4-amine 97%
2-Fluoro-N,N-dimethylpyridin-4-amine
Formula: C₇H₉FN₂
MW: 140,158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09031022
CAS Number: 849937-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR