6-Chloro-3-fluoropicolinaldehyde 98%

Supplier: Apollo Scientific

PC201351-1G PC201351-5G
APOSPC201351-1GEA 30.1 EUR
APOSPC201351-1G APOSPC201351-5G
6-Chloro-3-fluoropicolinaldehyde 98%
6-Chloro-3-fluoropicolinaldehyde
Formula: C₆H₃ClFNO
Storage Temperature: Ambient
MDL Number: MFCD04972390
CAS Number: 884494-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR